siRNA Details
Gene Symbol: ATPIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016311.4 | NP_057395.1 | 2, 3 | 266 | ||
GenBank mRNA | AB029042.1 | - | 2, 3 | 194 | ||
AF114836.1 | - | 352 | ||||
AK316600.1 | - | 2, 3 | 253 | |||
BC004955.1 | - | 2, 3 | 149 | 3625379 | ||
BC009677.1 | - | 2, 3 | 224 | 3877506 | ||
BG337531.1 | - | 2, 3 | 222 | |||
BP271519.1 | - | 2, 3 | 262 | |||
BT009849.1 | - | 2, 3 | 173 | |||
CR457097.1 | - | 2, 3 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
