siRNA Details
Gene Symbol: CXorf40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171907.2 | NP_001165378.1 | 5 | 1314 | ||
NM_001171908.2 | NP_001165379.1 | 5 | 1263 | |||
NM_001324274.1 | NP_001311203.1 | 5 | 1095 | |||
NM_001324275.1 | NP_001311204.1 | 5 | 1114 | |||
NM_001324277.1 | NP_001311206.1 | 5 | 1777 | |||
NM_001324278.1 | NP_001311207.1 | 5 | 1244 | |||
NM_001324280.1 | NP_001311209.1 | 5 | 1796 | |||
NM_178124.5 | NP_835225.2 | 5 | 1295 | |||
XM_005262349.4 | XP_005262406.2 | 4 | 1985 | |||
GenBank mRNA | AK055235.1 | - | 5 | 2214 | ||
AK291899.1 | - | 5 | 1221 | |||
AY074889.1 | - | 5 | 1257 | |||
BC065539.1 | - | 5 | 1220 | 6143134 | ||
L43576.1 | - | 2 | 491 |
Gene Symbol: CXorf40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013845.1 | NP_001013867.1 | 5 | 1231 | ||
XM_005274698.1 | XP_005274755.1 | 5 | 1602 | |||
XM_005274699.1 | XP_005274756.1 | 5 | 1634 | |||
XM_005274700.1 | XP_005274757.1 | 6 | 1780 | |||
XM_005274701.2 | XP_005274758.1 | 5 | 2143 | |||
XM_005274702.3 | XP_005274759.1 | 5 | 1337 | |||
XM_006724826.3 | XP_006724889.1 | 5 | 1356 | |||
XM_011531180.2 | XP_011529482.1 | 5 | 1447 | |||
XM_017029585.1 | XP_016885074.1 | 5 | 1653 | |||
XM_017029586.1 | XP_016885075.1 | 6 | 1799 | |||
XM_017029587.1 | XP_016885076.1 | 6 | 1831 | |||
XM_017029588.1 | XP_016885077.1 | 6 | 2340 | |||
XM_017029589.1 | XP_016885078.1 | 5 | 1437 | |||
GenBank mRNA | BC009523.1 | - | 3 | 837 | 3889701 | |
L43575.1 | - | 5 | 1157 | |||
L43578.1 | - | 4 | 959 | |||
U66042.1 | - | 5 | 1218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
