siRNA Details
Gene Symbol: CXorf40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013845.1 | NP_001013867.1 | 4 | 752 | ||
XM_005274698.1 | XP_005274755.1 | 4 | 1123 | |||
XM_005274699.1 | XP_005274756.1 | 4 | 1155 | |||
XM_005274700.1 | XP_005274757.1 | 5 | 1301 | |||
XM_005274701.2 | XP_005274758.1 | 4 | 1664 | |||
XM_005274702.3 | XP_005274759.1 | 4 | 858 | |||
XM_006724826.3 | XP_006724889.1 | 4 | 877 | |||
XM_011531180.2 | XP_011529482.1 | 4 | 968 | |||
XM_011531181.2 | XP_011529483.1 | 4 | 1104 | |||
XM_017029585.1 | XP_016885074.1 | 4 | 1174 | |||
XM_017029586.1 | XP_016885075.1 | 5 | 1320 | |||
XM_017029587.1 | XP_016885076.1 | 5 | 1352 | |||
XM_017029588.1 | XP_016885077.1 | 5 | 1861 | |||
XM_017029589.1 | XP_016885078.1 | 4 | 958 | |||
XM_017029590.1 | XP_016885079.1 | 4 | 1155 | |||
XM_017029591.1 | XP_016885080.1 | 4 | 1664 | |||
XM_017029592.1 | XP_016885081.1 | 4 | 1123 | |||
XM_017029593.1 | XP_016885082.1 | 5 | 1301 | |||
GenBank mRNA | BC009523.1 | - | 2 | 358 | 3889701 | |
L43575.1 | - | 4 | 678 | |||
L43578.1 | - | 3 | 480 | |||
U66042.1 | - | 4 | 739 |
Gene Symbol: CXorf40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171907.2 | NP_001165378.1 | 4 | 835 | ||
NM_001171908.2 | NP_001165379.1 | 4 | 784 | |||
NM_001171909.3 | NP_001165380.1 | 3 | 763 | |||
NM_001324274.1 | NP_001311203.1 | 4 | 616 | |||
NM_001324275.1 | NP_001311204.1 | 4 | 635 | |||
NM_001324276.1 | NP_001311205.1 | 4 | 835 | |||
NM_001324277.1 | NP_001311206.1 | 4 | 1298 | |||
NM_001324278.1 | NP_001311207.1 | 4 | 765 | |||
NM_001324279.1 | NP_001311208.1 | 4 | 784 | |||
NM_001324280.1 | NP_001311209.1 | 4 | 1317 | |||
NM_178124.5 | NP_835225.2 | 4 | 816 | |||
XM_005262349.4 | XP_005262406.2 | 3 | 1506 | |||
GenBank mRNA | AK055235.1 | - | 3 | 1823 | ||
AK291899.1 | - | 4 | 742 | |||
AK294153.1 | - | 4 | 811 | |||
AK300334.1 | - | 3 | 763 | |||
AY074889.1 | - | 4 | 778 | |||
BC065539.1 | - | 4 | 741 | 6143134 | ||
BC104930.1 | - | 4 | 684 | 8143933 | ||
BC104932.1 | - | 4 | 684 | 8143935 | ||
BC143660.1 | - | 4 | 665 | 9052174 | ||
BC143661.1 | - | 4 | 684 | 9052175 | ||
BC143662.1 | - | 4 | 632 | 9052176 | ||
BG831626.1 | - | 2 | 391 | |||
BI819316.1 | - | 4 | 528 | |||
DC339764.1 | - | 4 | 524 | |||
L43576.1 | - | 1 | 12 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
