siRNA Details
Gene Symbol: ACSM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308169.1 | NP_001295098.1 | 5 | 817 | ||
NM_001308172.1 | NP_001295101.1 | 6 | 1002 | |||
NM_001308954.1 | NP_001295883.1 | 7 | 1384 | |||
XM_017022923.1 | XP_016878412.1 | 7 | 977 | |||
XM_017022924.1 | XP_016878413.1 | 6 | 887 | |||
XM_017022925.1 | XP_016878414.1 | 5 | 774 | |||
XM_017022926.1 | XP_016878415.1 | 4 | 519 | |||
GenBank mRNA | AK055867.1 | - | 7 | 798 | ||
AK096039.1 | - | 7 | 1243 | |||
AK298359.1 | - | 5 | 706 | |||
AK298434.1 | - | 7 | 999 | |||
AK303774.1 | - | 5 | 702 | |||
BC125176.1 | - | 6 | 839 | 40121949 |
Gene Symbol: ACSM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105069.1 | NP_001098539.1 | 6 | 948 | ||
NM_182617.3 | NP_872423.3 | 7 | 983 | |||
XM_005255304.3 | XP_005255361.1 | 6 | 887 | |||
XM_011545824.2 | XP_011544126.1 | 5 | 769 | |||
XM_017023205.1 | XP_016878694.1 | 6 | 887 | |||
GenBank mRNA | AB073604.1 | - | 7 | 914 | ||
AK129944.1 | - | 6 | 887 | |||
AK289416.1 | - | 6 | 887 | |||
AK309601.1 | - | 6 | 948 | |||
AY160217.1 | - | 5 | 773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
