siRNA Details
Gene Symbol: ACSM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105069.1 | NP_001098539.1 | 13, 14 | 1797 | ||
NM_182617.3 | NP_872423.3 | 14, 15 | 1832 | |||
XM_011545824.2 | XP_011544126.1 | 12, 13 | 1618 | |||
GenBank mRNA | AB073604.1 | - | 14, 15 | 1763 | ||
AK129944.1 | - | 13, 14 | 1736 | |||
AY160217.1 | - | 12, 13 | 1622 |
Gene Symbol: ACSM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308169.1 | NP_001295098.1 | 12, 13 | 1666 | ||
NM_001308172.1 | NP_001295101.1 | 13, 14 | 1851 | |||
NM_001308954.1 | NP_001295883.1 | 14, 15 | 2233 | |||
XM_017022923.1 | XP_016878412.1 | 14, 15 | 1826 | |||
XM_017022926.1 | XP_016878415.1 | 11, 12 | 1368 | |||
GenBank mRNA | AK055867.1 | - | 14, 15 | 1647 | ||
AK091878.1 | - | 6, 7 | 1817 | |||
AK096039.1 | - | 14, 15 | 2092 | |||
AK298359.1 | - | 12, 13 | 1555 | |||
BC125176.1 | - | 13, 14 | 1688 | 40121949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
