siRNA Details
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317815.1 | NP_001304744.1 | 5 | 1140 | ||
NM_198853.2 | NP_942150.1 | 5 | 1201 | |||
XM_011516187.2 | XP_011514489.1 | 5 | 1109 | |||
XM_011516189.2 | XP_011514491.1 | 5 | 1422 | |||
GenBank mRNA | AF498999.1 | - | 5 | 1201 | ||
BC033871.1 | - | 5 | 1137 | 5201777 |
Gene Symbol: TRIM73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198924.3 | NP_944606.2 | 5 | 1201 | ||
GenBank mRNA | AF498998.1 | - | 5 | 1201 | ||
AK126632.1 | - | 2 | 3726 | |||
BC073926.1 | - | 5 | 1268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
