siRNA Details
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516187.2 | XP_011514489.1 | 1 | 74 | ||
XM_011516190.2 | XP_011514492.1 | 1 | 47 | |||
XM_017012197.1 | XP_016867686.1 | 8 | 1373 |
Gene Symbol: TRIM73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094396.1 | - | 1 | 344 | ||
BC073926.1 | - | 1 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
