siRNA Details
Gene Symbol: ZNF517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317936.1 | NP_001304865.1 | 3 | 181 | ||
NM_213605.2 | NP_998770.2 | 3 | 181 | |||
NR_134284.1 | 3 | 181 | ||||
XM_011517014.2 | XP_011515316.1 | 3 | 720 | |||
XM_011517015.2 | XP_011515317.1 | 3 | 162 | |||
GenBank mRNA | AK096527.1 | - | 3 | 720 | ||
AK097278.1 | - | 2 | 847 | |||
AK131440.1 | - | 3 | 181 | |||
BC126161.1 | - | 2 | 101 | 8991877 | ||
DB016003.1 | - | 3 | 181 | |||
DB165783.1 | - | 3 | 140 |
