siRNA Details
Gene Symbol: ZNF517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213605.2 | NP_998770.2 | 4 | 296 | ||
NR_134284.1 | 4 | 296 | ||||
NR_134285.1 | 3 | 169 | ||||
XM_011517014.2 | XP_011515316.1 | 4 | 835 | |||
XM_011517015.2 | XP_011515317.1 | 4 | 277 | |||
XM_011517017.2 | XP_011515319.1 | 3 | 694 | |||
GenBank mRNA | AK096527.1 | - | 4 | 835 | ||
AK097278.1 | - | 2 | 1319 | |||
AK131440.1 | - | 4 | 296 | |||
BC126161.1 | - | 3 | 216 | 8991877 | ||
BC143922.1 | - | 2 | 89 | |||
DB016003.1 | - | 4 | 296 | |||
DB152270.1 | - | 3 | 128 |
