siRNA Details
Gene Symbol: EPHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000534.1 | XP_016856023.1 | 7 | 1834 | ||
XM_017000535.1 | XP_016856024.1 | 7 | 1794 | |||
XM_017000536.1 | XP_016856025.1 | 6 | 1642 | |||
XM_017000537.1 | XP_016856026.1 | 7 | 1736 | |||
GenBank mRNA | AK296788.1 | - | 6 | 887 | ||
BC037166.2 | - | 7 | 1682 | 5090654 | ||
M59371.1 | - | 7 | 1677 |
