siRNA Details
Gene Symbol: AADAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286682.1 | NP_001273611.1 | 14 | 1389 | ||
NM_001286683.1 | NP_001273612.1 | 14 | 1462 | |||
NM_016228.3 | NP_057312.1 | 13 | 1595 | |||
NM_182662.1 | NP_872603.1 | 14 | 1377 | |||
XM_006714231.2 | XP_006714294.1 | 15 | 1519 | |||
XM_011532020.2 | XP_011530322.1 | 11 | 1152 | |||
GenBank mRNA | AF481738.1 | - | 13 | 1596 | ||
AK055952.1 | - | 14 | 1377 | |||
AK314729.1 | - | 14 | 1464 | |||
BC031068.1 | - | 14 | 1382 | 5286180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
