siRNA Details
Gene Symbol: IRAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025242.1 | NP_001020413.1 | 11 | 1604 | ||
NM_001569.3 | NP_001560.2 | 11 | 1604 | |||
XM_005274668.3 | XP_005274725.1 | 10 | 1626 | |||
GenBank mRNA | AF346607.1 | - | 11 | 1525 | ||
BC054000.1 | - | 11 | 1636 | 6164719 | ||
BE276817.1 | - | 2 | 107 | |||
L76191.1 | - | 11 | 1604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
