siRNA Details
Gene Symbol: PRLH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015893.1 | NP_056977.1 | 2 | 253 | ||
GenBank mRNA | AB015419.1 | - | 2 | 253 | ||
BC069081.1 | - | 2 | 253 | 7216895 | ||
BC069284.1 | - | 2 | 253 | 7262679 |
Gene Symbol: PIGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110061.1 | - | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
