siRNA Details
Gene Symbol: FGF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300812.1 | NP_001287741.1 | 3 | 390 | ||
NM_020637.1 | NP_065688.1 | 3 | 382 | |||
GenBank mRNA | AB021925.1 | - | 3 | 382 | ||
AY359084.1 | - | 3 | 388 | |||
BC137444.1 | - | 3 | 413 | 9021444 | ||
BC137445.1 | - | 3 | 413 | 9021445 | ||
BC144645.1 | - | 3 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
