siRNA Details
Gene Symbol: SCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021920.3 | NP_068739.1 | 3 | 282 | ||
XM_017018109.1 | XP_016873598.1 | 3 | 278 | |||
GenBank mRNA | HY019819.1 | - | 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
