siRNA Details
Gene Symbol: PCDH11Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278619.1 | NP_001265548.1 | 6 | 2787 | ||
NM_032971.2 | NP_116753.1 | 5 | 2618 | |||
NM_032972.2 | NP_116754.1 | 2 | 2872 | |||
NM_032973.2 | NP_116755.1 | 2 | 2872 | |||
XM_017030079.1 | XP_016885568.1 | 8 | 7149 | |||
XM_017030080.1 | XP_016885569.1 | 7 | 3101 | |||
XM_017030081.1 | XP_016885570.1 | 6 | 2871 | |||
XM_017030082.1 | XP_016885571.1 | 6 | 2684 | |||
XM_017030083.1 | XP_016885572.1 | 3 | 2448 | |||
GenBank mRNA | AF277053.1 | - | 6 | 2718 | ||
AF332216.1 | - | 2 | 2872 | |||
AF332217.1 | - | 2 | 2872 | |||
AJ276803.1 | - | 5 | 2618 | |||
AJ564960.1 | - | 1 | 1502 | |||
AJ564961.1 | - | 1 | 1502 | |||
AJ564962.1 | - | 1 | 1502 | |||
AJ564963.1 | - | 1 | 1502 | |||
AK289676.1 | - | 5 | 2409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
