siRNA Details
Gene Symbol: AVP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000490.4 | NP_000481.2 | 2 | 228 | ||
XM_011529267.1 | XP_011527569.1 | 3 | 1758 | |||
GenBank mRNA | AF031476.1 | - | 2 | 196 | ||
BC126196.1 | - | 2 | 205 | 8991912 | ||
BC126224.1 | - | 2 | 205 | 8991940 | ||
BG715518.1 | - | 2 | 233 | |||
KJ534777.1 | - | 2 | 178 | |||
KJ534778.1 | - | 2 | 178 | |||
M25647.1 | - | 2 | 201 | |||
X03172.1 | - | 2 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
