siRNA Details
Gene Symbol: H2BFWT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002916.4 | NP_001002916.3 | 1, 2 | 529 | ||
GenBank mRNA | AY283370.1 | - | 1, 2 | 502 | ||
BC038109.2 | - | 1, 2 | 526 | 5266336 | ||
BC118604.1 | - | 1, 2 | 522 | 40109007 | ||
BC121816.1 | - | 1, 2 | 522 | 40109006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
