siRNA Details
Gene Symbol: CYP4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000896.2 | NP_000887.2 | 6 | 608 | ||
NM_001199208.1 | NP_001186137.1 | 6 | 608 | |||
NM_001199209.1 | NP_001186138.1 | 6 | 592 | |||
XM_005259911.4 | XP_005259968.1 | 4 | 536 | |||
XM_011528014.2 | XP_011526316.1 | 4 | 385 | |||
XM_017026814.1 | XP_016882303.1 | 5 | 686 | |||
XM_017026815.1 | XP_016882304.1 | 5 | 691 | |||
GenBank mRNA | AB002454.1 | - | 6 | 608 | ||
AK296761.1 | - | 6 | 527 | |||
AK296802.1 | - | 5 | 698 | |||
AK298675.1 | - | 6 | 588 | |||
AK304200.1 | - | 6 | 605 | |||
AK316136.1 | - | 6 | 592 | |||
BC136299.1 | - | 6 | 600 | 9020286 | ||
D12620.1 | - | 6 | 592 | |||
D12621.1 | - | 6 | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
