siRNA Details
Gene Symbol: C19orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018111.2 | NP_060581.2 | 1 | 278 | ||
GenBank mRNA | AK001352.1 | - | 1 | 278 | ||
BC051004.1 | - | 1 | 283 | 6483373 | ||
BC069669.1 | - | 1 | 278 | 7262644 | ||
BC069693.1 | - | 1 | 278 | 7262656 | ||
BC069712.1 | - | 1 | 278 | 7262573 | ||
BC069750.1 | - | 1 | 278 | 7262585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
