siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 7 | 1280 | |||
NR_027776.2 | 6 | 1177 | ||||
GenBank mRNA | AK309660.1 | - | 5 | 1036 | ||
D38439.1 | - | 6 | 1056 | |||
D38501.1 | - | 6 | 1056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
