siRNA Details
Gene Symbol: GATC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176818.2 | NP_789788.1 | 2 | 246 | ||
NR_033684.1 | 2 | 246 | ||||
GenBank mRNA | AK094319.1 | - | 2 | 240 | ||
BC034962.1 | - | 2 | 217 | |||
BC047778.1 | - | 2 | 217 | |||
BC064523.1 | - | 2 | 217 | |||
BC107145.1 | - | 2 | 203 | 40031296 | ||
BC107146.1 | - | 2 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
