siRNA Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001016.3 | NP_001007.2 | 1 | 145 | ||
GenBank mRNA | AK311826.1 | - | 3 | 140 | ||
BC002550.1 | - | 1230 | ||||
BC017321.2 | - | 1 | 128 | 4565075 | ||
BC071930.1 | - | 1 | 124 | 6295016 | ||
BC095424.1 | - | 1 | 135 | 30792687 | ||
BQ276878.1 | - | 3 | 148 | |||
X53505.1 | - | 3 | 142 |
Gene Symbol: SNORA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096147.1 | - | 1 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
