siRNA Details
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002157.2 | NP_002148.1 | 4 | 748 | ||
GenBank mRNA | BC023518.2 | - | 4 | 422 | 4304224 | |
BG188453.1 | - | 3 | 309 | |||
U07550.1 | - | 4 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
