siRNA Details
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144822.1 | NP_001138294.1 | 4, 5 | 828 | ||
NM_001779.2 | NP_001770.1 | 4, 5 | 828 | |||
XM_017002869.1 | XP_016858358.1 | 4, 5 | 828 | |||
GenBank mRNA | AK291980.1 | - | 4, 5 | 761 | ||
BT019816.1 | - | 4, 5 | 707 | |||
BT019817.1 | - | 4, 5 | 707 | |||
D28586.1 | - | 2, 3 | 359 | |||
Y00636.1 | - | 4, 5 | 716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
