siRNA Details
Gene Symbol: IL17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013278.3 | NP_037410.1 | 2, 3 | 386 | ||
GenBank mRNA | AF152099.1 | - | 2, 3 | 386 | ||
AY358471.1 | - | 2, 3 | 386 | |||
BC069152.1 | - | 1, 2 | 337 | 7217012 | ||
BC101835.1 | - | 2, 3 | 381 | 8069341 | ||
BC112196.1 | - | 2, 3 | 381 | 8327664 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
