siRNA Details
Gene Symbol: NCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145466.1 | NP_001138938.1 | 3 | 745 | ||
NM_001145467.1 | NP_001138939.1 | 3 | 745 | |||
NM_147130.2 | NP_667341.1 | 3 | 745 | |||
XM_006715049.3 | XP_006715112.1 | 4 | 691 | |||
XM_011514459.2 | XP_011512761.1 | 4 | 709 | |||
GenBank mRNA | AB055881.1 | - | 3 | 481 | ||
AF031136.1 | - | 3 | 745 | |||
AF031137.1 | - | 3 | 745 | |||
AF031138.1 | - | 3 | 745 | |||
AJ223153.1 | - | 3 | 544 | |||
BC018752.1 | - | 3 | 709 | 4849421 | ||
BC052582.1 | - | 3 | 689 | 5934651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
