siRNA Details
Gene Symbol: MT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005953.3 | NP_005944.1 | 3 | 359 | ||
GenBank mRNA | BC007034.1 | - | 1 | 344 | 4051220 | |
BC070289.1 | - | 1 | 338 | 4051053 | ||
V00594.1 | - | 3 | 326 | |||
X97260.1 | - | 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
