siRNA Details
Gene Symbol: SYNGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004209.5 | NP_004200.2 | 2 | 300 | ||
GenBank mRNA | AJ002309.2 | - | 2 | 146 | ||
AK313894.1 | - | 2 | 267 | |||
BC009568.1 | - | 2 | 246 | 3932502 | ||
BC014087.2 | - | 2 | 220 | 4334996 | ||
BG699055.1 | - | 2 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
