siRNA Details
Gene Symbol: NXNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161625.1 | NP_001155097.1 | 1 | 540 | ||
NM_145283.2 | NP_660326.2 | 1 | 540 | |||
XM_005251727.3 | XP_005251784.1 | 1 | 546 | |||
XM_011518275.2 | XP_011516577.1 | 1 | 543 | |||
XM_011518276.2 | XP_011516578.1 | 1 | 553 | |||
GenBank mRNA | BC022521.1 | - | 1 | 536 | 4793993 | |
BI517442.1 | - | 1 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
