siRNA Details
Gene Symbol: NXNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161625.1 | NP_001155097.1 | 1 | 590 | ||
NM_145283.2 | NP_660326.2 | 1 | 590 | |||
XM_005251727.3 | XP_005251784.1 | 1 | 596 | |||
XM_011518275.2 | XP_011516577.1 | 1 | 593 | |||
XM_011518276.2 | XP_011516578.1 | 1 | 603 | |||
GenBank mRNA | BC022521.1 | - | 1 | 586 | 4793993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
