siRNA Details
Gene Symbol: FUT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002034.2 | NP_002025.2 | 2 | 382 | ||
GenBank mRNA | BC140905.1 | - | 2 | 378 | 9056717 | |
U27329.1 | - | 2 | 382 | |||
U27330.1 | - | 2 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
