siRNA Details
Gene Symbol: ULBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025217.3 | NP_079493.1 | 5 | 1075 | ||
XM_017011321.1 | XP_016866810.1 | 4 | 934 | |||
GenBank mRNA | AB052906.1 | - | 5 | 1013 | ||
AX772820.1 | - | 4 | 865 | |||
AY358665.1 | - | 5 | 948 | |||
BC034689.1 | - | 5 | 970 | 4747126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
