siRNA Details
Gene Symbol: ULBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025217.3 | NP_079493.1 | 5 | 1355 | ||
XM_017011321.1 | XP_016866810.1 | 4 | 1214 | |||
GenBank mRNA | AB052906.1 | - | 5 | 1293 | ||
AX772820.1 | - | 4 | 1145 | |||
AY358665.1 | - | 5 | 1228 | |||
BC034689.1 | - | 5 | 1250 | 4747126 |
Gene Symbol: RAET1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535486.2 | XP_011533788.1 | 5 | 1354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
