siRNA Details
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002306.3 | NP_002297.2 | 5 | 617 | ||
NR_003225.2 | 3 | 1573 | ||||
XM_011536759.2 | XP_011535061.2 | 6 | 607 | |||
GenBank mRNA | AB006780.1 | - | 5 | 582 | ||
AB209391.1 | - | 4 | 4660 | |||
AF266280.1 | - | 3 | 900 | |||
AK314929.1 | - | 5 | 616 | |||
BC001120.2 | - | 5 | 553 | 3050135 | ||
BC053667.1 | - | 5 | 568 | 6148401 | ||
BC068068.1 | - | 5 | 563 | 6428428 | ||
BQ645629.1 | - | 681 | ||||
BX161455.1 | - | 3 | 1368 | |||
BX641090.1 | - | 3 | 1574 | |||
CD518618.1 | - | 5 | 617 | |||
CR456897.1 | - | 4 | 529 | |||
CR542097.1 | - | 4 | 529 | |||
M35368.1 | - | 5 | 551 | |||
M36682.1 | - | 5 | 562 | |||
M57710.1 | - | 4 | 547 | |||
M64303.1 | - | 4 | 547 | |||
S59012.1 | - | 4 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
