siRNA Details
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002306.3 | NP_002297.2 | 6 | 765 | ||
NR_003225.2 | 4 | 1721 | ||||
XM_011536759.2 | XP_011535061.2 | 7 | 755 | |||
GenBank mRNA | AB006780.1 | - | 6 | 730 | ||
AB209391.1 | - | 5 | 4808 | |||
AF266280.1 | - | 4 | 1048 | |||
AK314929.1 | - | 6 | 764 | |||
BC001120.2 | - | 6 | 701 | 3050135 | ||
BC053667.1 | - | 6 | 716 | 6148401 | ||
BC068068.1 | - | 6 | 711 | 6428428 | ||
BX161455.1 | - | 4 | 1516 | |||
BX641090.1 | - | 4 | 1722 | |||
CR456897.1 | - | 5 | 677 | |||
CR542097.1 | - | 5 | 677 | |||
M35368.1 | - | 6 | 699 | |||
M36682.1 | - | 6 | 710 | |||
M57710.1 | - | 5 | 695 | |||
M64303.1 | - | 5 | 695 | |||
S59012.1 | - | 5 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
