siRNA Details
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177388.1 | NP_001170859.1 | 6 | 638 | ||
NM_002306.3 | NP_002297.2 | 6 | 911 | |||
NR_003225.2 | 4 | 1867 | ||||
XM_011536759.2 | XP_011535061.2 | 7 | 901 | |||
GenBank mRNA | AB006780.1 | - | 6 | 876 | ||
AB209391.1 | - | 5 | 4954 | |||
BG332134.1 | - | 6 | 595 | |||
BX161455.1 | - | 4 | 1662 | |||
BX641090.1 | - | 4 | 1868 | |||
M35368.1 | - | 6 | 847 | |||
M57710.1 | - | 5 | 843 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
