siRNA Details
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018052.3 | NP_060522.3 | 7, 8 | 1072 | ||
XM_005256038.3 | XP_005256095.1 | 7, 8 | 1083 | |||
XM_005256041.3 | XP_005256098.2 | 7, 8 | 1083 | |||
XM_011523223.2 | XP_011521525.1 | 7, 8 | 1083 | |||
XM_011523224.2 | XP_011521526.1 | 7, 8 | 1083 | |||
XM_011523225.2 | XP_011521527.1 | 7, 8 | 1083 | |||
XM_017023441.1 | XP_016878930.1 | 7, 8 | 1083 | |||
XM_017023442.1 | XP_016878931.1 | 7, 8 | 1083 | |||
GenBank mRNA | AK025479.1 | - | 1, 2 | 105 | ||
AK056433.1 | - | 7, 8 | 1082 | |||
AK297564.1 | - | 6, 7 | 851 | |||
AK307766.1 | - | 6, 7 | 922 | |||
BC007214.1 | - | 2, 3 | 194 | |||
BC125108.1 | - | 7, 8 | 1004 | 40118772 | ||
BC125109.1 | - | 7, 8 | 1004 | 40118777 | ||
CR457343.1 | - | 1, 2 | 40 | |||
U25801.1 | - | 1, 2 | 85 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
