siRNA Details
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018052.3 | NP_060522.3 | 7 | 1024 | ||
XM_005256038.3 | XP_005256095.1 | 7 | 1035 | |||
XM_005256041.3 | XP_005256098.2 | 7 | 1035 | |||
XM_011523223.2 | XP_011521525.1 | 7 | 1035 | |||
XM_011523224.2 | XP_011521526.1 | 7 | 1035 | |||
XM_011523225.2 | XP_011521527.1 | 7 | 1035 | |||
XM_017023441.1 | XP_016878930.1 | 7 | 1035 | |||
XM_017023442.1 | XP_016878931.1 | 7 | 1035 | |||
GenBank mRNA | AK056433.1 | - | 7 | 1034 | ||
AK297564.1 | - | 6 | 803 | |||
AK307766.1 | - | 6 | 874 | |||
BC007214.1 | - | 2 | 146 | |||
BC125108.1 | - | 7 | 956 | 40118772 | ||
BC125109.1 | - | 7 | 956 | 40118777 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146216, AK140191, AK171883, AK046826, AK171577, and others.
Gene Ontology Categories:
