siRNA Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145312.1 | NP_001138784.1 | 3, 4 | 581 | ||
NM_005240.1 | NP_005231.1 | 3, 4 | 379 | |||
XM_006711210.2 | XP_006711273.1 | 3, 4 | 562 | |||
GenBank mRNA | AF218540.1 | - | 2, 3 | 292 | ||
AK304789.1 | - | 3, 4 | 385 | |||
AK316274.1 | - | 3, 4 | 581 | |||
BC022868.1 | - | 3, 4 | 379 | |||
L16464.1 | - | 3, 4 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
