siRNA Details
Gene Symbol: PPP1R3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006242.3 | NP_006233.1 | 1 | 949 | ||
GenBank mRNA | BC047502.1 | - | 1 | 949 | 5312727 | |
BC074860.2 | - | 1 | 630 | 30915227 | ||
BC074861.2 | - | 1 | 630 | 30915464 | ||
Y18206.1 | - | 1 | 859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
