siRNA Details
Gene Symbol: CHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253387.1 | NP_001240316.1 | 9 | 1605 | ||
NM_001253388.1 | NP_001240317.1 | 8 | 1105 | |||
NM_006614.3 | NP_006605.2 | 10 | 1653 | |||
XM_006712938.1 | XP_006713001.1 | 10 | 1264 | |||
XM_006712939.3 | XP_006713002.1 | 9 | 1581 | |||
XM_006712940.3 | XP_006713003.1 | 9 | 1495 | |||
XM_011533292.1 | XP_011531594.1 | 10 | 1634 | |||
XM_011533294.1 | XP_011531596.1 | 9 | 1216 | |||
XM_011533295.1 | XP_011531597.1 | 9 | 1586 | |||
XM_011533296.1 | XP_011531598.1 | 10 | 1264 | |||
XM_017005566.1 | XP_016861055.1 | 11 | 6158 | |||
XM_017005567.1 | XP_016861056.1 | 9 | 1366 | |||
XM_017005568.1 | XP_016861057.1 | 9 | 1173 | |||
XM_017005569.1 | XP_016861058.1 | 8 | 1533 | |||
XM_017005570.1 | XP_016861059.1 | 8 | 1447 | |||
XM_017005571.1 | XP_016861060.1 | 8 | 1318 | |||
XM_017005572.1 | XP_016861061.1 | 10 | 1661 | |||
XM_017005573.1 | XP_016861062.1 | 9 | 1581 | |||
GenBank mRNA | AB209329.1 | - | 9 | 1430 | ||
AF002246.1 | - | 10 | 1282 | |||
AK126878.1 | - | 9 | 1584 | |||
BC104918.1 | - | 7 | 994 | 8143921 | ||
BC143496.1 | - | 7 | 991 | 9052004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
