siRNA Details
Gene Symbol: AGO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017629.3 | NP_060099.2 | 10 | 1342 | ||
XM_005270578.3 | XP_005270635.1 | 10 | 1227 | |||
XM_005270579.3 | XP_005270636.1 | 9 | 1035 | |||
XM_011540883.2 | XP_011539185.1 | 7 | 831 | |||
XM_011540884.2 | XP_011539186.1 | 7 | 694 | |||
GenBank mRNA | AB046787.1 | - | 10 | 1342 | ||
AL831908.1 | - | 9 | 1129 | |||
BC152450.1 | - | 10 | 1342 | 8862582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
