siRNA Details
Gene Symbol: PLEKHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197026.1 | NP_001183955.1 | 3 | 678 | ||
NM_001197027.1 | NP_001183956.1 | 3 | 678 | |||
NM_032639.3 | NP_116028.1 | 3 | 678 | |||
XM_005249897.2 | XP_005249954.1 | 3 | 626 | |||
XM_011515591.2 | XP_011513893.1 | 3 | 626 | |||
XM_011515592.2 | XP_011513894.1 | 3 | 626 | |||
XM_011515594.1 | XP_011513896.1 | 3 | 626 | |||
XM_011515595.1 | XP_011513897.1 | 3 | 626 | |||
XM_011515596.2 | XP_011513898.1 | 2 | 161 | |||
XM_017012744.1 | XP_016868233.1 | 3 | 626 | |||
XM_017012745.1 | XP_016868234.1 | 3 | 626 | |||
XM_017012746.1 | XP_016868235.1 | 3 | 626 | |||
XM_017012747.1 | XP_016868236.1 | 3 | 626 | |||
XM_017012748.1 | XP_016868237.1 | 2 | 219 | |||
XM_017012749.1 | XP_016868238.1 | 3 | 266 | |||
GenBank mRNA | AF308300.1 | - | 3 | 354 | ||
AK023180.1 | - | 3 | 598 | |||
AK294857.1 | - | 3 | 627 | |||
BC002838.2 | - | 3 | 367 | 3635744 | ||
BC053990.1 | - | 3 | 614 | 5922853 |
Gene Symbol: PLEKHA8P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037144.1 | 3 | 465 | |||
GenBank mRNA | AF103731.1 | - | 3 | 465 | ||
AF380162.1 | - | 1 | 294 | |||
BC028914.1 | - | 1 | 306 | |||
BC063575.1 | - | 3 | 346 | 3881751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
