siRNA Details
Gene Symbol: HLA-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243042.1 | NP_001229971.1 | 2 | 359 | ||
NM_002117.5 | NP_002108.4 | 2 | 359 | |||
GenBank mRNA | AJ010749.1 | - | 2 | 294 | ||
AJ440716.2 | - | 1 | 221 | |||
AK024836.1 | - | 2 | 316 | |||
AK130592.1 | - | 2 | 255 | |||
AK301283.1 | - | 2 | 423 | |||
AM237285.2 | - | 2 | 294 | |||
BC007814.2 | - | 2 | 292 | |||
BC010542.1 | - | 2 | 292 | |||
BC033293.1 | - | 2 | 301 | 4780246 | ||
CA395932.1 | - | 2 | 301 | |||
D12471.1 | - | 2 | 294 | |||
D38526.1 | - | 2 | 294 | |||
D49552.1 | - | 2 | 294 | |||
D49819.1 | - | 2 | 294 | |||
D64146.1 | - | 2 | 294 | |||
D64152.1 | - | 2 | 294 | |||
D83741.1 | - | 1 | 221 | |||
M11886.1 | - | 1 | 222 | |||
M21963.1 | - | 1 | 171 | |||
M28207.1 | - | 1 | 66 | |||
U06695.1 | - | 2 | 294 | |||
U06696.1 | - | 2 | 294 | |||
U09853.1 | - | 2 | 294 | |||
X70856.1 | - | 2 | 298 | |||
X82122.1 | - | 2 | 294 | |||
X83394.1 | - | 2 | 315 | |||
X97321.1 | - | 2 | 294 | |||
Z46810.1 | - | 2 | 294 |
Gene Symbol: HLA-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG514457.1 | - | 2 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
