siRNA Details
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006382.3 | NP_006373.2 | 12 | 2222 | ||
GenBank mRNA | BC143291.1 | - | 5 | 498 | ||
BC171723.1 | - | 3 | 260 | |||
U43383.1 | - | 5 | 516 | |||
U65652.1 | - | 5 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
