siRNA Details
Gene Symbol: CCM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080625.3 | NP_542192.2 | 4 | 383 | ||
XM_006723706.3 | XP_006723769.1 | 4 | 446 | |||
XM_011528566.1 | XP_011526868.1 | 4 | 446 | |||
XM_011528567.2 | XP_011526869.1 | 4 | 446 | |||
XM_011528568.2 | XP_011526870.1 | 5 | 572 | |||
XM_011528569.1 | XP_011526871.1 | 4 | 446 | |||
GenBank mRNA | AK057090.1 | - | 4 | 380 | ||
BC030254.1 | - | 4 | 375 | 5182496 | ||
CA406781.1 | - | 4 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
