siRNA Details
Gene Symbol: CCM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080625.3 | NP_542192.2 | 4, 5 | 482 | ||
XM_006723706.3 | XP_006723769.1 | 4, 5 | 545 | |||
XM_011528566.1 | XP_011526868.1 | 4, 5 | 545 | |||
XM_011528567.2 | XP_011526869.1 | 4, 5 | 545 | |||
XM_011528568.2 | XP_011526870.1 | 5, 6 | 671 | |||
XM_011528569.1 | XP_011526871.1 | 4, 5 | 545 | |||
GenBank mRNA | AK057090.1 | - | 4, 5 | 479 | ||
BC030254.1 | - | 4, 5 | 474 | 5182496 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
