siRNA Details
Gene Symbol: MAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015329.3 | NP_056144.3 | 16, 17 | 1592 | ||
XM_005259837.3 | XP_005259894.2 | 16, 17 | 1581 | |||
XM_005259838.3 | XP_005259895.3 | 14, 15 | 1643 | |||
XM_011527844.2 | XP_011526146.1 | 16, 17 | 1997 | |||
XM_017026539.1 | XP_016882028.1 | 14, 15 | 1639 | |||
GenBank mRNA | AB020699.1 | - | 16, 17 | 1578 | ||
AK001349.1 | - | 4, 5 | 309 | |||
AK025896.1 | - | 14, 15 | 1244 | |||
AK126227.1 | - | 3, 4 | 1484 | |||
AL110250.1 | - | 5, 6 | 367 | |||
BC010222.1 | - | 5, 6 | 378 | |||
BC063863.1 | - | 5, 6 | 725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
