siRNA Details
Gene Symbol: MAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015329.3 | NP_056144.3 | 3 | 356 | ||
XM_005259837.3 | XP_005259894.2 | 3 | 348 | |||
XM_006722711.3 | XP_006722774.2 | 3 | 348 | |||
XM_011527844.2 | XP_011526146.1 | 3 | 761 | |||
GenBank mRNA | AB020699.1 | - | 3 | 342 | ||
BY795974.2 | - | 3 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
